Learn More
Grainger QMARK Electric Wall Heater, Programmable Touchscreen, 208V, 1800-4000 W
Catalog No. 502769678
Encompass_Preferred
This electric fan-forced architectural wall heater provides precise heating solutions for commercial environments. Featuring a digital touch screen thermostat, it offers 5/2-day programmability with four daily periods and proportional heat control to maintain room temperatures within 1.5°F of the set point. The robust unit includes a 20-gauge steel louvered grille and is designed for either recessed installation between wall studs or surface mounting.
  • Proportional heating automatically adjusts wattage output for optimal comfort
  • Built-in building management system (BMS) connection for dry contact control
  • Integrated manual-reset safety limit control with red overheat warning light
  • Totally enclosed fan motor with five-bladed aluminum fan
  • Concealed electronic disconnect switch
  • 5/2-day programmable thermostat with lockout security feature
Catalog No. 50-276-9678 Supplier Grainger Supplier No. 13R108
This electric fan-forced architectural wall heater provides precise heating solutions for commercial environments. Featuring a digital touch screen thermostat, it offers 5/2-day programmability with four daily periods and proportional heat control to maintain room temperatures within 1.5°F of the set point. The robust unit includes a 20-gauge steel louvered grille and is designed for either recessed installation between wall studs or surface mounting.
  • Proportional heating automatically adjusts wattage output for optimal comfort
  • Built-in building management system (BMS) connection for dry contact control
  • Integrated manual-reset safety limit control with red overheat warning light
  • Totally enclosed fan motor with five-bladed aluminum fan
  • Concealed electronic disconnect switch
  • 5/2-day programmable thermostat with lockout security feature