Learn More
Grainger Qmark Recessed Electric Wall-Mount Heater, 4000 Watts, 240 Volts, 1 ea
Catalog No. 502769375
Encompass_Preferred
The SmartSeries fan-forced wall heater is designed for seamless integration with building climate control systems, featuring a 4,000-watt output suitable for large areas up to 400 sq. ft. This recessed unit offers precise temperature management through an integrated LED touchscreen-enabled programmable thermostat. It is compatible with Building Management Systems (BMS) and utilizes proportional heating to automatically adjust wattage for optimal comfort and energy efficiency.
  • 5/2-day programmable thermostat with four daily time periods
  • Proportional heat output maintains temperatures within 1.5°F of set point
  • BMS compatible for centralized monitoring and control
  • Includes fan-only ventilation mode and security lockout feature
  • Three-piece construction for recessed installation
  • Equipped with manual-reset safety limit control and power on/off disconnect switch
Catalog No. 50-276-9375 Supplier Grainger Supplier No. 13R109
The SmartSeries fan-forced wall heater is designed for seamless integration with building climate control systems, featuring a 4,000-watt output suitable for large areas up to 400 sq. ft. This recessed unit offers precise temperature management through an integrated LED touchscreen-enabled programmable thermostat. It is compatible with Building Management Systems (BMS) and utilizes proportional heating to automatically adjust wattage for optimal comfort and energy efficiency.
  • 5/2-day programmable thermostat with four daily time periods
  • Proportional heat output maintains temperatures within 1.5°F of set point
  • BMS compatible for centralized monitoring and control
  • Includes fan-only ventilation mode and security lockout feature
  • Three-piece construction for recessed installation
  • Equipped with manual-reset safety limit control and power on/off disconnect switch