Learn More
Chemglass Life Sciences Large Extraction Apparatus Complete, Soxhlet, 55/50, 24/40
SDP
Catalog No. CG136903
Encompass_Preferred
Extraction Apparatus, Soxhlet, Large, 55/50, 24/40. Similar to CG-1368, but supplied with a round bottom instead of a flat bottom flask. All specifications are the same as CG-1368.
Catalog No. CG136903 Supplier Chemglass Life Sciences Supplier No. CG136903
Extraction Apparatus, Soxhlet, Large, 55/50, 24/40. Similar to CG-1368, but supplied with a round bottom instead of a flat bottom flask. All specifications are the same as CG-1368.