Learn More
Nordson Medical PharmaLok Large Tube Clamp, wall thickness 1/8 (3.2mm) to 1/4 (6.4mm), max OD 2.5 (63.5mm), Glass filled white nylon
Catalog No. 502083649
Encompass_Preferred
harmaLok Large Tube Clamp, wall thickness 1/8 (3.2mm) to 1/4 (6.4mm), max OD 2.5 (63.5mm), Glass filled white nylon 100/cs
Catalog No. 50-208-3649 Supplier Nordson Medical Supplier No. CPT110U71
harmaLok Large Tube Clamp, wall thickness 1/8 (3.2mm) to 1/4 (6.4mm), max OD 2.5 (63.5mm), Glass filled white nylon 100/cs