Learn More
Nanotemper Technologies Inc Large Volume Labeling Kit RED-NHS 2nd Generation
Catalog No. NC3781759
Encompass
This Large Volume Protein Labeling Kit RED-NHS 2nd Generation kit increases data quality and enables affinity studies with previously challenging targets by generating higher binding amplitudes and improved signal-to-noise ratios. It contains all required reagents for labeling and dye removal. The dye carries a reactive NHS-ester group that reacts with primary amines (lysine residues) to form a covalent bond. The kit is optimized for proteins with a molecular weight higher than 5 kDa and concentrations between 2 and 20 µM. RED dyes are suited for Monolith instruments with a RED detector (Nano and Pico) and Dianthus instruments. Contents require storage at 4 °C and -20 °C.
Catalog No. NC3781759 Supplier Nanotemper Technologies Inc Supplier No. NTL111
This Large Volume Protein Labeling Kit RED-NHS 2nd Generation kit increases data quality and enables affinity studies with previously challenging targets by generating higher binding amplitudes and improved signal-to-noise ratios. It contains all required reagents for labeling and dye removal. The dye carries a reactive NHS-ester group that reacts with primary amines (lysine residues) to form a covalent bond. The kit is optimized for proteins with a molecular weight higher than 5 kDa and concentrations between 2 and 20 µM. RED dyes are suited for Monolith instruments with a RED detector (Nano and Pico) and Dianthus instruments. Contents require storage at 4 °C and -20 °C.