Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 29124MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: PP13/1162. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b. Positive Control: K-562 cells. Placenta or Spleen. Cellular Staining: Cytoplasmic ;Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Galectin-13 protein fragment (aa23-134). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 16kDa (monomer) ;32kDa (homodimer).Chromosome: 19q13.1. Gene Symbol: LGALS13. Gene ID: 29124. Ensemble: ENSG00000105198. Uniprot: XP_016882204.
Catalog No. 50-188-4756