Learn More
Aviva Systems Biology Corp LGALS13 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP42376_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LGALS13. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human LGALS13. Alternate names: GAL13, PLAC8, PP13ApplicationWB. Species reactivity: Human
Catalog No. 50-284-997