Learn More
Aviva Systems Biology Corp LGALS14 Antibody - N-terminal region (ARP42549_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42549_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LGALS14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LGALS14. Alternate names: CLC2, MGC22235, PPL13ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58261