Learn More
Aviva Systems Biology Corp LGALS3 Antibody - N-terminal region (ARP54688_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54688_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LGALS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LGALS3. Alternate names: CBP35, GAL3, GALBP, GALIG, LGALS2, MAC2, L31ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61286