Learn More
Aviva Systems Biology Corp LGALS7 Antibody - middle region (ARP64143_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64143_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LGALS7. It was validated on Western Blot. . Alternate names: GAL7, LGALS7AApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-185