Learn More
Aviva Systems Biology Corp LGALS8 Antibody - C-terminal region (ARP53656_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53656_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LGALS8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LGALS8. Alternate names: Gal-8, PCTA-1, PCTA1, Po66-CBPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61081