Learn More
Medchemexpress LLC LGD 3303 | 917891-35-1 | MFCD17167063 | ≥98.0% | 342.7 g/mol | C16H14ClF3N2O | 5 MG
SDP
Catalog No. 5000244993
Encompass_Preferred
LGD 3303 is a non-steroidal selective androgen receptor modulator (SARM) supplied as a solid research reagent for receptor binding, reporter gene, and preclinical in vivo studies. It selectively binds the human androgen receptor (reported Ki = 0.9 nM) and is provided at research-grade purity for in vitro and in vivo experiments.
  • Selective androgen receptor agonist (reported Ki = 0.9 nM for human receptor).
  • High purity (≥98%).
  • Molecular weight 342.7 g/mol; chemical formula C16H14ClF3N2O.
  • Supplied in small research pack sizes (for example, 5 mg).
  • Intended for research use only; not for human consumption.
Catalog No. 50-002-44993 Supplier Medchemexpress LLC Supplier No. HY1035765MG
LGD 3303 is a non-steroidal selective androgen receptor modulator (SARM) supplied as a solid research reagent for receptor binding, reporter gene, and preclinical in vivo studies. It selectively binds the human androgen receptor (reported Ki = 0.9 nM) and is provided at research-grade purity for in vitro and in vivo experiments.
  • Selective androgen receptor agonist (reported Ki = 0.9 nM for human receptor).
  • High purity (≥98%).
  • Molecular weight 342.7 g/mol; chemical formula C16H14ClF3N2O.
  • Supplied in small research pack sizes (for example, 5 mg).
  • Intended for research use only; not for human consumption.