Learn More
Medchemexpress LLC LGD-6972 | 1207989-09-0 | MFCD28411370 | 98.0% | 702.90 g/mol | C43H46N2O5S | 25MG
SDP
Catalog No. 5000235053
Encompass_Preferred
LGD-6972 is an orally active, selective glucagon receptor (GCGR) antagonist used for research into glucose regulation and type 2 diabetes. It inhibits GCGR signaling to reduce glucagon-stimulated cAMP and hepatic glucose production; reported molecular formula C43H46N2O5S, molecular weight 702.9 g/mol, CAS 1207989-09-0, and typical supplied purity is 98.0%.
  • Selective oral GCGR antagonist for glucose regulation research.
  • Reduces glucagon-stimulated cAMP and hepatic glucose production.
  • Demonstrated glucose-lowering activity in preclinical and clinical studies.
  • Available in multiple salt forms and small-scale pack sizes for laboratory use.
  • High purity suitable for analytical and biological assays.
Catalog No. 50-002-35053 Supplier Medchemexpress LLC Supplier No. HY1252525MG
LGD-6972 is an orally active, selective glucagon receptor (GCGR) antagonist used for research into glucose regulation and type 2 diabetes. It inhibits GCGR signaling to reduce glucagon-stimulated cAMP and hepatic glucose production; reported molecular formula C43H46N2O5S, molecular weight 702.9 g/mol, CAS 1207989-09-0, and typical supplied purity is 98.0%.
  • Selective oral GCGR antagonist for glucose regulation research.
  • Reduces glucagon-stimulated cAMP and hepatic glucose production.
  • Demonstrated glucose-lowering activity in preclinical and clinical studies.
  • Available in multiple salt forms and small-scale pack sizes for laboratory use.
  • High purity suitable for analytical and biological assays.