Learn More
Cayman Chemical LGD 6972amonIum salt 10mg
SDP
Catalog No. 502945076
Encompass_Preferred
A GCGR antagonist; reduces glucagon-induced hyperglycemia in wild-type mice; reduces blood glucose levels in mouse models of diabetes
Catalog No. 50-294-5076 Supplier Cayman Chemical Supplier No. 4096110 MG
A GCGR antagonist; reduces glucagon-induced hyperglycemia in wild-type mice; reduces blood glucose levels in mouse models of diabetes