Learn More
Aviva Systems Biology Corp LGI1 Antibody - N-terminal region (ARP61478_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61478_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LGI1. It was validated on Western Blot. . Alternate names: EPITEMPIN, EPT, ETL1, IB1099, ADLTE, ADPAEFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-392