Learn More
Selleck Chemical LLC LGK-974
SDP
Catalog No. 5000001354
Encompass_Preferred
LGK-974 (NVP-LGK974 WNT974) is a potent and specific PORCN inhibitor and inhibits Wnt signaling with IC50 of 0 4 nM in TM3 cells Phase 1
Catalog No. 50-000-01354 Supplier Selleck Chemical LLC Supplier No. S71431G
Add to Cart
Add to Cart
Request a Quote
LGK-974 (NVP-LGK974 WNT974) is a potent and specific PORCN inhibitor and inhibits Wnt signaling with IC50 of 0 4 nM in TM3 cells Phase 1