Learn More
Medchemexpress LLC HY-17545 10mg Medchemexpress, LGK974 CAS:1243244-14-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1754510MG
Encompass_Preferred
Medchemexpress, HY-17545 10mg LGK974 CAS:1243244-14-5 LGK974 (WNT974) is an orally bioavailable and specific Porcupine (PORCN) inhibitor with an IC50 of 0.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9243