Learn More
BIOGEMS INTERNATIONAL INC LGK974 5mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 12414545MG
Encompass
LGK974 is a highly potent inhibitor of Porcupine (PORCN), a Wnt-specific acyltransferase. LGK974 is orally bioavailable and inhibits WNT signaling in vitro and in vivo. It is reported to exhibit anti-tumor properties without increasing cytotoxicity.
Catalog No. 50-152-9700