Learn More
Abcam LGK974, 5MG
Catalog No. 502671248
Encompass_Preferred
MW 396.4 Da, Purity >=98%. LGK-974 is a potent and specific inhibitor of Porcupine (porcn), a Wnt-specific acyltransferase (IC₅₀ = ~0.4 nM). GK974 potently inhibits Wnt signaling, has strong efficacy in rodent tumor models, and is well-tolerated. In addition, head and neck cancer cell lines with loss-of-function mutations in the Notch signaling pathway have a high response rate to LGK974.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1248 Supplier Abcam Supplier No. AB2854045MG
MW 396.4 Da, Purity >=98%. LGK-974 is a potent and specific inhibitor of Porcupine (porcn), a Wnt-specific acyltransferase (IC₅₀ = ~0.4 nM). GK974 potently inhibits Wnt signaling, has strong efficacy in rodent tumor models, and is well-tolerated. In addition, head and neck cancer cell lines with loss-of-function mutations in the Notch signaling pathway have a high response rate to LGK974.
The product is subject to the following: Abcam Restricted Use Statement