Learn More
Selleck Chemical LLC LGK-974 5mg 1243244-14-5 NVP-LGK974, WNT974
SDP
Supplier:  Selleck Chemical LLC S71435MG
Encompass_Preferred
LGK-974 (NVP-LGK974, WNT974) is a potent and specific PORCN inhibitor, and inhibits Wnt signaling with IC50 of 0.4 nM in TM3 cells. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0671899