Learn More
Cayman Chemical L-Glutathione, reduced, Purity: ≥95%, CAS: 70-18-8, 5 g
SDP
Catalog No. 502890851
Encompass_Preferred
Nucleophilic co-substrate to glutathione transferases and electron donor to glutathione peroxidases; involved in amino acid transport and maintenance of protein sulfhydryl reduction status
Catalog No. 50-289-0851 Supplier Cayman Chemical Supplier No. 100074615 G
Nucleophilic co-substrate to glutathione transferases and electron donor to glutathione peroxidases; involved in amino acid transport and maintenance of protein sulfhydryl reduction status