Learn More
Cayman Chemical L-Glutathione, reduced, Purity: ≥95%, CAS: 70-18-8, 10 g
SDP
Catalog No. 502888181
Encompass_Preferred
Nucleophilic co-substrate to glutathione transferases and electron donor to glutathione peroxidases; involved in amino acid transport and maintenance of protein sulfhydryl reduction status
Catalog No. 50-288-8181 Supplier Cayman Chemical Supplier No. 1000746110 G
Nucleophilic co-substrate to glutathione transferases and electron donor to glutathione peroxidases; involved in amino acid transport and maintenance of protein sulfhydryl reduction status