Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LGP2 protein, human (HEK293, FLAG) | >90.0% | 10UG
SDP

Catalog No. 5000174906
Encompass_Preferred

Recombinant human LGP2 protein expressed in HEK293 cells with a C-terminal FLAG tag. The protein regulates RIG-I and MDA5-mediated antiviral signaling and is supplied as a purified, buffered solution intended for research applications. Provided in small microgram quantities, the product includes storage and handling recommendations to preserve activity and integrity.

  • Expressed in HEK293 cells with C-terminal FLAG tag.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Formulated in 50 mM HEPES, pH 7.5; 200 mM NaCl; 20% glycerol; 1 mM DTT.
  • Available in 10 μg, 50 μg, and 100 μg sizes.
  • Store at -80°C; stable at -20°C for short-term; avoid repeated freeze-thaw cycles.
  • Molecular weight approximately 77.6 kDa (coverage M1-D678).

Catalog No. 50-001-74906 Supplier Medchemexpress LLC Supplier No. HYP70199810UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human LGP2 protein expressed in HEK293 cells with a C-terminal FLAG tag. The protein regulates RIG-I and MDA5-mediated antiviral signaling and is supplied as a purified, buffered solution intended for research applications. Provided in small microgram quantities, the product includes storage and handling recommendations to preserve activity and integrity.

  • Expressed in HEK293 cells with C-terminal FLAG tag.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Formulated in 50 mM HEPES, pH 7.5; 200 mM NaCl; 20% glycerol; 1 mM DTT.
  • Available in 10 μg, 50 μg, and 100 μg sizes.
  • Store at -80°C; stable at -20°C for short-term; avoid repeated freeze-thaw cycles.
  • Molecular weight approximately 77.6 kDa (coverage M1-D678).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.