Learn More
Novus Biologicals™ Lgr5/GPR49 Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 8549 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NLS1235. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 |
| Gene Symbol | LGR5 |
| Quantity | 0.1 mg |
| Research Category | Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.