Learn More
Aviva Systems Biology Corp LGTN Antibody - middle region (ARP40690_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40690_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LGTN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LGTN. Alternate names: HCA56, LGTNApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57588