Learn More
Cayman Chemical β-L-Gulopyranosyl-caldarchaetidyl-glycerol, Purity: ≥95%, CAS: 416880-23-4, 50 mg
SDP
Catalog No. 502937864
Encompass_Preferred
The main polar lipid of the archaeon T. acidophilum; cellular levels of β-L-gulopyranosyl-caldarchaetidyl-glycerol in T. acidophilum HO-62 positively and negatively correlate with increases in culture pH and temperature, respectively; has been used as a standard for the quantification of polar lipids extracted from T. acidophilum HO-62
Catalog No. 50-293-7864 Supplier Cayman Chemical Supplier No. 2720350 MG
The main polar lipid of the archaeon T. acidophilum; cellular levels of β-L-gulopyranosyl-caldarchaetidyl-glycerol in T. acidophilum HO-62 positively and negatively correlate with increases in culture pH and temperature, respectively; has been used as a standard for the quantification of polar lipids extracted from T. acidophilum HO-62