Learn More
Medchemexpress LLC HY-15605 10mg , LGX818 Encorafenib CAS:1269440-17-6 Purity:98%
SDP
Catalog No. 501494034
Encompass_Preferred
Medchemexpress, HY-15605 10mg LGX818 Encorafenib CAS:1269440-17-6 Formula:C22H27ClFN7O4S IC50: 0.3 nM (BRaf V600E ) Purity:98% LGX818 is an orally available mutated BRaf V600E inhibitor ( IC 50 =0.3 nM) with potential antineoplastic activity. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4034 Supplier Medchemexpress LLC Supplier No. HY1560510MG
Medchemexpress, HY-15605 10mg LGX818 Encorafenib CAS:1269440-17-6 Formula:C22H27ClFN7O4S IC50: 0.3 nM (BRaf V600E ) Purity:98% LGX818 is an orally available mutated BRaf V600E inhibitor ( IC 50 =0.3 nM) with potential antineoplastic activity. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.