Learn More
Enzo Life Sciences LH ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1070001
Encompass_Preferred
Highly sensitive LH ELISA kit enabling detection of LH in serum, plasma and tissue culture media in just 2 hours. The LH ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 hours. Alternative Name: Luteinizing hormone. Lutropin. Sensitivity: 5.2 mIU/ml (1.2-280 mIU/ml). Assay Time: 2 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human, Rat. Contents: Microtiter Plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Tracer, Conjugate, Substrate, and Stop Solution
Catalog No. NC1501468