Learn More
Bellco Glass, Inc. LH Pitch Blade Imp 2 3/4in,7L
SDP
Catalog No. 501946443
Encompass_Preferred
SST pitched blade impeller with left hand oriented Blades 2.75" od.Assemble one or more impellers onto 3L – 15L drive shaft for mammalian cell culture.. MADE IN THE USA
Catalog No. 50-194-6443 Supplier Bellco Glass, Inc. Supplier No. 790900034
SST pitched blade impeller with left hand oriented Blades 2.75" od.Assemble one or more impellers onto 3L – 15L drive shaft for mammalian cell culture.. MADE IN THE USA