Learn More
Bellco Glass, Inc. LH Pitch Blade Imp 3 1/2,15L
SDP
Catalog No. 501946489
Encompass_Preferred
Stainless steal pitch blade impeller with left hand oriented blades 2’OD.Assemble one or more impellers into 3L – 15L drive shaft for mammaliancell culture.. LH Pitch Blade Impeller 3 1/2" 15L. MADE IN THE USA
Catalog No. 50-194-6489 Supplier Bellco Glass, Inc. Supplier No. 790915106
Stainless steal pitch blade impeller with left hand oriented blades 2’OD.Assemble one or more impellers into 3L – 15L drive shaft for mammaliancell culture.. LH Pitch Blade Impeller 3 1/2" 15L. MADE IN THE USA