Learn More
BioVendor, LLC LH (S-type) Rat ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC 29488501
Encompass
LH (S-type) Rat ELISA (96 wells). Sandwich ELISA immunoassay kit for quantification of luteinizing hormone in rat serum or plasma. SAMPLE REQUIREMENT: 10 l/well. CALIBRATION RANGE: 0.31310 ng/ml. LIMIT OF DETECTION: 31.3 pg/ml. OTHER NAMES: lutropin, lutrophin. SUMMARY: Luteinizing hormone (LH) is produced and released in the anterior pituitary gland. This hormone is considered a gonadotrophic hormone because of its role in controlling the function of ovaries in females and testes in males, which are known as the gonads. Manufactured by FUJIFILM Wako Shibayagi Corporation, Japan. OTHER NAMES: anti-Ovalbumin.
Catalog No. 50-423-9