Learn More
Echelon Biosciences Research Labs LH-RH [D-Lys6] - 5 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 550465MG
Encompass_Preferred
LH-RH [D-Lys6] - 5mg , Glp-His-Trp-Ser-Tyr-D-Lys-Leu-Arg-Pro-Gly-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8613