Learn More
Eurofins Discoverx Luteinizing Hormone Releasing Hormone (LH-RH), Human, 5 mg. (92-1206)
Supplier:  Eurofins Discoverx 921206
Encompass
Luteinizing Hormone Releasing Hormone (LH-RH, GnRH) is a hypothalamic peptide hormone that stimulates the production and release of follicle-stimulating hormone (FSH) and luteinizing hormone (LH) from the anterior pituitary, subsequently initiating and maintaining reproductive organ functions. Agonist of the gonadotropin-releasing hormone receptor (GnRHR). Provided as 5mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9733
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.