Learn More
CPC Scientific Glp-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific LHRH001A
Encompass
A hypothalamic neuropeptide that regulates reproductive functions by controlling the synthesis and release of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) from the anterior pituitary gland.ONE-LETTER SEQUENCE: Glp-HWSYGLRPG-NH2MOLECULAR FORMULA: C55H75N17O13MOLECULAR WEIGHT:1182.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [71447-49-9], RESEARCH AREA: HormonalREFERENCES: J.E. Rivier et al., Life Sci. 23, 869 (1978); D. Tilemans et al., Endocrinology, 130, 887 (1992)
Catalog No. 50-210-9645