Learn More
Drg International Inc DRG LH-Serum ELISA, 96 Well Kit
Supplier:  Drg International Inc EIA1289
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of the luteinizing hormone (LH) in serum. It is composed of two non covalently associated dissimilar amino acid chains, alpha and beta. The alpha chain is similar to that found in human thyroid-stimulating hormone (TSH), follicle stimulating hormone (FSH), and human chorionic gonadotropin (hCG). The difference between these hormones lie in the amino acid composition of their beta subunits, which account for their immunological differentiation. The DRG LH-Serum ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site on a LH molecule.
Catalog No. 50-187-1253