Learn More
enQuire Bioreagents LH, alpha anti-Human Monoclonal [LHa/756], Size: 100ug
Supplier:  enQuire Bioreagents 1081MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: LHa/756. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Anterior Pituitary. Cellular Staining: Cytoplasmic, secreted. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length hLH alpha protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 13kDa.Chromosome: 6q12-q21. Gene Symbol: LHA. Gene ID: 1081. Ensemble: ENSG00000135346. Uniprot: NP_000726.
Catalog No. 50-188-4590