Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3972MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: LHb/1214. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Anterior Pituitary. Cellular Staining: Cytoplasmic, secreted. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant beta sub-unit of human LH. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 22kDa.Chromosome: 19q13.32. Gene Symbol: LHB. Gene ID: 3972. Ensemble: ENSG00000104826. Uniprot: XP_011525277.
Catalog No. 50-188-4910