Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ LHC-8 Medium (1X), without gentamicin
GREENER_CHOICE

Catalog No. 12679015
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-679-015 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-679-015 Supplier Gibco™ Supplier No. 12679015
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

LHC-8 Medium (1X), without gentamicin

LHC serum-free media are the most widely used products for culture of bronchial epithelial cells. This is a supplemented medium. For maximum flexibility and convenience, we also offer LHC basal medium.

Applications:

Engineered specifically for the growth of bronchial epithelial cells such as IB3-1, S9, and C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), and BBE (bovine bronchial epithelial cells) for asthma, allergy, lung cancer, cystic fibrosis, pulmonary, and esophageal cancer research without further supplements.

Performance and quality testing:

Performance tested in a growth assay using BEAS-2B cells.

Reference(s):

Pawliczak R., et. al. (2001) J Biol Chem 276, 44613. Zhao Y.L., et. al. (2000) Carcinogenesis 21, 2005. Liu X., et. al. (1998) Am J Physiol 274, L58. Hei T.K., et. al. (1997) Environ Health Perspect 105, 1085. Lechner J.F., and LaVeck, M.A. (1985) J Tissue Cult Methods 9, 43.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line B3-1, S9, C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), BBE (bovine bronchial epithelial cells)
Cell Type Epithelial Cells
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type LHC-8 Medium
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Without Additives No Gentamicin
Quantity 500 mL
Shipping Condition Dry Ice
Included Antibiotics No Gentamicin Sulfate
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.