Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ LHC Basal Medium (1X)
GREENER_CHOICE

Catalog No. 12677027
Encompass_Preferred
Change view
Click to view available options
Quantity:
1,000 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-677-027 1,000 mL
12-677-019 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-677-027 Supplier Gibco™ Supplier No. 12677027
  • $73.50
    $67.65 / Each of 1

    Save $5.85

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LHC Basal Medium (1X)

LHC serum-free media are the most widely used products for culture of bronchial epithelial cells. For maximum flexibility and convenience, we offer these products with or without supplementation.

LHC Basal Medium (1X), liquid

  • Applications: Formulated for the growth of bronchial epithelial cells with further supplements.
  • Recommended storage conditions: 2°C to 8°C.
  • Shelf life: 12 months

LHC-8 Medium and LHC-9 Medium

  • Applications: Engineered specifically for the growth of bronchial epithelial cells such as IB3-1, S9, and C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), and BBE (bovine bronchial epithelial cells) for asthma, allergy, lung cancer, cystic fibrosis, pulmonary, and esophageal cancer research without further supplements.
  • Performance and quality testing: Performance tested in a growth assay using BEAS-2B cells.
  • Recommended storage conditions: -5°C to -20°C.

Reference(s):

  1. Pawliczak R., et. al. (2001) J Biol Chem 276, 44613.
  2. Zhao Y.L., et. al. (2000) Carcinogenesis 21, 2005.
  3. Liu X., et. al. (1998) Am J Physiol 274, L58.
  4. Hei T.K., et. al. (1997) Environ Health Perspect 105, 1085.
  5. Lechner J.F., and LaVeck, M.A. (1985) J Tissue Cult Methods 9, 43.
TRUSTED_SUSTAINABILITY

Specifications

Cell Line Bronchial epithelial cells, IB3-1, S9, C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), BBE (bovine bronchial epithelial cells)
Cell Type Epithelial Cell (Bronchial)
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type LHC Basal Medium
Sterility Sterile-filtered
With Additives Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate
Quantity 1,000 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.