Learn More
Eurofins Discoverx cAMP Hunter LHCGR Bioassay Kit, 2-Plate. (95-0106Y2-00137)
Catalog No. 502003106
Encompass
The cAMP Hunter CHO-K1 LHCGR Bioassay Kit provides a robust, and highly sensitive functional, cell-based assay to measure drug potency and detect neutralizing antibodies.. The bioassay kit contains all the materials needed for a complete assay, including cells, detection reagents, cell plating reagent, positive control agonist, and assay plates. The ready-to-use cryopreserved cells have been manufactured to ensure assay reproducibility and faster implementation from characterization to lot release. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-3106 Supplier Eurofins Discoverx Supplier No. 950106Y200137
The cAMP Hunter CHO-K1 LHCGR Bioassay Kit provides a robust, and highly sensitive functional, cell-based assay to measure drug potency and detect neutralizing antibodies.. The bioassay kit contains all the materials needed for a complete assay, including cells, detection reagents, cell plating reagent, positive control agonist, and assay plates. The ready-to-use cryopreserved cells have been manufactured to ensure assay reproducibility and faster implementation from characterization to lot release. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.