Learn More
Cayman Chemical LHomopropargyl GlycInhydr 50mg
SDP
Catalog No. 502951958
Encompass_Preferred
A reactive methionine analog that contains an alkyne moiety that is readily inserted into newly-synthesized proteins; can be labeled or captured through click chemistry
Catalog No. 50-295-1958 Supplier Cayman Chemical Supplier No. 1178550 MG
A reactive methionine analog that contains an alkyne moiety that is readily inserted into newly-synthesized proteins; can be labeled or captured through click chemistry