Learn More
Cayman Chemical L-Homoserine lactone (hydrochloride), Purity: ≥95%, CAS: 2185-03-7, 250 mg
SDP
Catalog No. 502992171
Encompass_Preferred
A synthetic intermediate; has been used in the synthesis of organoselenium chemistry intermediates and analogs of bacterial quorum sensing signaling molecules
Catalog No. 50-299-2171 Supplier Cayman Chemical Supplier No. 31492250 MG
A synthetic intermediate; has been used in the synthesis of organoselenium chemistry intermediates and analogs of bacterial quorum sensing signaling molecules