Learn More
Aviva Systems Biology Corp LHPP Antibody - middle region (ARP57605_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57605_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LHPP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LHPP. Alternate names: MGC117251, MGC142189, MGC142191, HDHD2BApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62982