Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LHPP Human His-SUM 5ug
SDP

Catalog No. 5000196336
Encompass_Preferred

Recombinant human LHPP (phospholysine phosphohistidine inorganic pyrophosphate phosphatase) is supplied lyophilized with an N-terminal SUMO and 6xHis tag, produced in Escherichia coli and purified for research use as a small-quantity 5 μg vial. This high-purity protein is formulated for stability and intended for biochemical characterization and assay development.

  • N-terminal SUMO and 6xHis tags facilitate detection and purification.
  • Expressed in Escherichia coli for recombinant production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized from PBS with 6% trehalose (pH 7.4) for enhanced stability.
  • Recommended storage at -20°C; aliquot and store at -80°C for extended storage.
  • Provided as a 5 μg quantity suitable for biochemical assays.

Catalog No. 50-001-96336 Supplier Medchemexpress LLC Supplier No. HYP716325UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human LHPP (phospholysine phosphohistidine inorganic pyrophosphate phosphatase) is supplied lyophilized with an N-terminal SUMO and 6xHis tag, produced in Escherichia coli and purified for research use as a small-quantity 5 μg vial. This high-purity protein is formulated for stability and intended for biochemical characterization and assay development.

  • N-terminal SUMO and 6xHis tags facilitate detection and purification.
  • Expressed in Escherichia coli for recombinant production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized from PBS with 6% trehalose (pH 7.4) for enhanced stability.
  • Recommended storage at -20°C; aliquot and store at -80°C for extended storage.
  • Provided as a 5 μg quantity suitable for biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.