Learn More
Bachem LHRH Acetate
SDP
Supplier:  Bachem 4033013.0025
Encompass_Preferred
LHRH Acetate, Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2, 33515-09-2, C55H75N17O13, Mr 1182.31, 25mg. GnRH, Gonadorelin, Luliberin, LH-RH, human. A hypothalamic neuropeptide which stimulates the release of LH and FSH. CAS Number (gonadorelin acetate): 34973-08-5.
Catalog No. 50-259-794