Learn More
CPC Scientific Pyr-His-Trp-Ser-Tyr-Gly-Leu-Gln-Pro-Gly-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific LHRH012A
Encompass
Chicken GnRH I and its isoform GnRH II were detected in the chicken hypothalamus.ONE-LETTER SEQUENCE: Pyr-HWSYGLQPG-NH2MOLECULAR FORMULA: C54H71N15O14MOLECULAR WEIGHT:1154.25STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [47922-48-5], RESEARCH AREA: HormonalREFERENCES: J.A.King and R.P.Millar, J. Biol. Chem., 257, 10729 (1982); J.A.King et al., J. Endocrinol., 117, 43 (1988)
Catalog No. 50-210-9659