Learn More
CPC Scientific Pyr-His-Trp-Ser-His-Gly-Trp-Tyr-Pro-Gly-NH2
(trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific LHRH020A
Encompass
The GnRH II sequence pEHWSHGWYPG amide, first detected in chicken, is highly conserved in vertebrates.ONE-LETTER SEQUENCE: Pyr-HWSHGWYPG-NH2MOLECULAR FORMULA: C60H69N17O13MOLECULAR WEIGHT:1236.32STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [91097-16-4], RESEARCH AREA: HormonalREFERENCES: K.Miyamoto et al., Proc. Natl. Acad. Sci. USA, 81, 3874 (1984); J.A.King et al., J. Endocrinol., 117, 43 (1988); J.F.Powell et al., Regul. Peptides, 57, 43 (1995); J.F.Powell et al., Regul. Peptides, 68, 189 (1997); A.Chen et al., Endocrinology, 142, 830 (2001); C.Gründker et al., Eur. J. Endocrinol., 151, 141 (2004); F.Tanriverdi et al., Clin. Exp. Immunol., 142, 103 (2005); D.K.Barnett et al., Endocrinology, 147, 615 (2006); I.-S.Hong et al., J. Clin. Endocrinol. Metab., 93, 3179 (2008)
Catalog No. 50-210-9671