Learn More
NABERTHERM INC These compact high-temperature furnaces have a variety of advantages. High-quality materials and ease of operation make these furnaces all-rounders in research.
Supplier:  NABERTHERM INC LHB257WN4240
Encompass_Preferred
High temperatures furnaces with Tmax 1600°C, 1750°C or 1800°C; non-classified fiber insulation; perfectly suited for the sintering of technical ceramics, such as zirconium oxide dental bridges; molybdenum disilicide heating elements; textured stainless steel housing
Catalog No. NC2332314
Add to Cart
Request a Quote