Learn More
Aviva Systems Biology Corp LHX3 Antibody - middle region (ARP33643_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33643_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LHX3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human LHX3. Alternate names: LIM3, CPHD3, M2-LHX3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-55106