Learn More
Aviva Systems Biology Corp LHX3 Antibody - middle region (ARP32443_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32443_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LHX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LHX3. Alternate names: DKFZp762A2013, M2-LHX3, LIM3, CPHD3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-54590